accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NaH3CO

CAS RN 141-52-6

IUPAC Name: sodium ethanolate
Synonyms: sodium ethoxide ethoxysodium NaOEt sodium ethylate caustic alcohol
Molecular Weight (g/mol): 68.05
Molecular Formula: C2H5NaO
InChi Key: QDRKDTQENPPHOJ-UHFFFAOYSA-N
SMILES: [Na+].CC[O-]
Molecular Weight (g/mol) 
  • (5)
  • (3)
  • (9)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
Percent Purity 
  • (3)
  • (2)
  • (6)
Grade 
  • (6)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (8)
  • (6)
  • (3)
  • (5)
  • (3)
  • (9)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)
  • (2)
  • (6)
  • (6)
  • (2)
  • (2)