accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NaOO

CAS RN 141-53-7

IUPAC Name: sodium formate
Synonyms: sodium formate sodium formiate formic acid, NA salt formic acid, sodium salt formic acid, sodium salt (1:1)
Molecular Weight (g/mol): 68.01
Molecular Formula: CHNaO2
InChi Key: HLBBKKJFGFRGMU-UHFFFAOYSA-M
SMILES: [Na+].[O-]C=O
Molecular Weight (g/mol) 
  • (23)
  • (5)
Quantity 
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (7)
  • (1)
Percent Purity 
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (3)
  • (1)
Grade 
  • (1)
  • (8)
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
  • (1)
  • (1)
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (2)
  • (2)
  • (3)
  • (3)
  • (2)
  • (5)
  • (5)
  • (2)
  • (1)
  • (23)
  • (5)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (7)
  • (1)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (8)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (3)
  • (2)
  • (5)
  • (2)
  • (2)