missing translation for 'onlineSavingsMsg'
Learn More

Sodium formate, Extra Pure, SLR, Fisher Chemical

Product Code. 10224590 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
500 g
Packaging:
Plastic powder jar
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10224590 500 g Plastic powder jar 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10224590 Supplier Fisher Chemical Supplier No. S/4080/53

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Boiling Point 360°C
Melting Point 258°C
CAS 141-53-7
Color White
pH 7 to 8.5
Physical Form Powder/Solid
Quantity 500 g
Molecular Formula CHNaO2
MDL Number 13101
Synonym sodium formate, formic acid, sodium salt, salachlor, formic acid sodium salt, formic acid, na salt, sodium formate, refined, sodium formiate, mravencan sodny czech, ccris 1037, hsdb 744
InChI Key HLBBKKJFGFRGMU-UHFFFAOYSA-M
SMILES C(=O)[O-].[Na+]
IUPAC Name sodium;formate
Molecular Weight (g/mol) 68.007
PubChem CID 2723810
ChEBI CHEBI:62965
Formula Weight 68.01g/mol
Packaging Plastic powder jar
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.