accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3CH3CH3O

CAS RN 141-79-7

IUPAC Name: 4-methylpent-3-en-2-one
Synonyms: mesityl oxide 4-methylpent-3-EN-2-one
Molecular Weight (g/mol): 98.15
Molecular Formula: C6H10O
InChi Key: SHOJXDKTYKFBRD-UHFFFAOYSA-N
SMILES: CC(C)=CC(C)=O
Molecular Weight (g/mol) 
  • (6)
  • (5)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
Boiling Point 
  • (3)
  • (4)
Percent Purity 
  • (4)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results