accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 141-97-9

IUPAC Name: ethyl 3-oxobutanoate
Synonyms: ethyl acetoacetate acetoctan ethylnaty ethylester kyseliny acetoctove EAA acetoacetic acid, ethyl ester, calcium complex ( ethyl acetylacetonate ethylacetacetat active acetylacetate acetoacetic ester active acetyl acetate
Molecular Weight (g/mol): 130.14
Molecular Formula: C6H10O3
InChi Key: XYIBRDXRRQCHLP-UHFFFAOYSA-N
SMILES: CCOC(=O)CC(C)=O
Molecular Weight (g/mol) 
  • (9)
  • (6)
Quantity 
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
Boiling Point 
  • (5)
  • (4)
  • (3)
Physical Form 
  • (12)
Percent Purity 
  • (4)
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (9)
  • (6)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (5)
  • (4)
  • (3)
  • (12)
  • (4)
  • (3)
  • (3)
  • (3)
  • (3)