accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 14104-20-2

AgFBFFF

CAS RN 14104-20-2

IUPAC Name: silver(1+) tetrafluoroboranuide
Synonyms: silver tetrafluoroborate
Molecular Weight (g/mol): 194.67
Molecular Formula: AgBF4
InChi Key: CCAVYRRHZLAMDJ-UHFFFAOYSA-N
SMILES: [Ag+].F[B-](F)(F)F
Quantity 
  • (2)
  • (3)
  • (1)
  • (2)
Physical Form 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (3)