missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Silver Tetrafluoroborate, TRC
Specifications
Specifications
| Chemical Name or Material | Silver Tetrafluoroborate |
| CAS | 14104-20-2 |
| Quantity | 10 g |
| Molecular Formula | AgBF4 |
| Synonym | Silver Fluoborate','Silver Tetrafluoroborate, Boron Silver Fluoride (BAgF4), Silver Borofluoride, Silver Borofluoride AgBF4, Silver Fluoroborate, Silver Fluoroborate (AgBF4), Silver Tetrafluoroborate, Silver(1+) Tetrafluoroborate, Silver(1+) Tetrafluoroborate, Silver(I) Tetrafluoroborate |
| InChI Formula | InChI=1S/Ag.BF4/c;2-1(3,4)5/q+1;-1 |
| SMILES | F[B](F)(F)(F)[Ag] |
| IUPAC Name | argentio(tetrafluoro)boron |
| Molecular Weight (g/mol) | 194.67 |
| Formula Weight | 193.908 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?