accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 141556-45-8

CAS RN 141556-45-8

IUPAC Name: 1,3-bis(2,4,6-trimethylphenyl)-3H-1λ⁵-imidazol-1-ylium
Synonyms: 1,3-bis(2,4,6-trimethylphenyl)-1λ⁵-imidazol-1-ylium
Molecular Weight (g/mol): 305.44
Molecular Formula: C21H25N2
InChi Key: CSRGPERAJKNQMM-UHFFFAOYSA-N
SMILES: CC1=CC(C)=C(N2C=C[N+](=C2)C2=C(C)C=C(C)C=C2C)C(C)=C1
Molecular Weight (g/mol) 
  • (3)
  • (4)
Quantity 
  • (3)
  • (1)
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (3)
  • (3)
  • (4)
  • (3)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)