missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dimesitylimidazolium Chloride, TRC

Product Code. 30377928
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
500 mg
missing translation for 'unitSize'
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30377928 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 30377928 missing translation for 'mfr' Toronto Research Chemicals missing translation for 'supplierNo' TRCD460683500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1,3-Dimesitylimidazolium Chloride
CAS 141556-45-8
Molecular Formula C21H25ClN2
Quantity 500 mg
Synonym Imes Hydrochloride, 1,3-Dimesityl-1H-imidazolium Chloride, 1,3-Dihydro-1,3-dimesityl-2H-imidazol-2-ylidene Monohydrochloride, 1,3-Bis(mesityl)imidazolium Chloride, 1,3-Bis(2,4,6-trimethylphenyl)imidazolium Chloride, 1,3-Bis(2,4,6-trimethylphenyl)-1H-imidazolium Chloride
InChI Formula InChI=1S/C21H25N2.ClH/c1-14-9-16(3)20(17(4)10-14)22-7-8-23(13-22)21-18(5)11-15(2)12-19(21)6;/h7-13H,1-6H3;1H/q+1;/p-1
SMILES [Cl-].Cc1cc(C)c(c(C)c1)[n+]2ccn(c2)c3c(C)cc(C)cc3C
IUPAC Name 1,3-bis(2,4,6-trimethylphenyl)imidazol-1-ium;chloride
Molecular Weight (g/mol) 340.9
Formula Weight 340.1706
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.