accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

ONH

CAS RN 142-08-5

IUPAC Name: 1,2-dihydropyridin-2-one
Synonyms: 2(1H)-pyridinone
Molecular Weight (g/mol): 95.1
Molecular Formula: C5H5NO
InChi Key: UBQKCCHYAOITMY-UHFFFAOYSA-N
SMILES: O=C1NC=CC=C1
Boiling Point 
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)