missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxypyridine, 98%

Product Code. 11451878
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11451878 500 g 500g
11431878 25 g 25g
11441878 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11451878 Supplier Thermo Scientific Alfa Aesar Supplier No. A14522.36
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 142-08-5 | C5H5NO | 95.10 g/mol

2-Hydroxypyridine is a bifunctional catalyst for a wide variety of acylation reactions, catalyst for generating β-oxopropyl carbonates from cyclic carbonates and alcohols and in the aminolysis of a polyglutamate. It is used in peptide synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Hydroxypyridine is a bifunctional catalyst for a wide variety of acylation reactions, catalyst for generating β-oxopropyl carbonates from cyclic carbonates and alcohols and in the aminolysis of a polyglutamate. It is used in peptide synthesis.

Solubility
Soluble in water 450 g/L @ 20°C and ethanol, slightly soluble in benzene and ether.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 142-08-5
Molecular Formula C5H5NO
Molecular Weight (g/mol) 95.10
MDL Number MFCD00006268
InChI Key UBQKCCHYAOITMY-UHFFFAOYSA-N
Synonym 2-hydroxypyridine, pyridin-2-ol, 2-pyridinol, 2-pyridone, pyridin-2 1h-one, 2 1h-pyridinone, 2-pyridinone, 2-oxopyridine, 2 1h-pyridone, alpha-pyridone
PubChem CID 8871
ChEBI CHEBI:16540
SMILES O=C1NC=CC=C1

Specifications

Melting Point 102°C to 111°C
Density 1.39
Boiling Point 280°C to 281°C
Flash Point 210°C (410°F)
Odor Recognizable
Quantity 500 g
UN Number UN2811
Beilstein 105786
Solubility Information Soluble in water 450g/L @ 20°C and ethanol,slightly soluble in benzene and ether.
IUPAC Name 1H-pyridin-2-one
Formula Weight 95.1
Percent Purity 98%
Chemical Name or Material 2-Hydroxypyridine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.