accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 142-71-2

CuCH3OOCH3OO

CAS RN 142-71-2

IUPAC Name: copper(2+) diacetate
Synonyms: cupric acetate copper(II) acetate
Molecular Weight (g/mol): 181.63
Molecular Formula: C4H6CuO4
InChi Key: OPQARKPSCNTWTJ-UHFFFAOYSA-L
SMILES: [Cu++].CC([O-])=O.CC([O-])=O
Molecular Weight (g/mol) 
  • (2)
  • (9)
  • (3)
Physical Form 
  • (3)
  • (9)
  • (2)
Quantity 
  • (1)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (3)
Grade 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (9)
  • (2)
  • (9)
  • (3)
  • (3)
  • (9)
  • (2)
  • (1)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (3)
  • (3)