missing translation for 'onlineSavingsMsg'
Learn More

Copper(II) Acetate, Anhydrous, TRC

Product Code. 30352224
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30352224 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30352224 Supplier Toronto Research Chemicals Supplier No. TRCC68535550G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Copper(II) Acetate, Anhydrous
CAS 142-71-2
Molecular Formula 2(C2H3O2)- . (Cu)2+
Quantity 50 g
Synonym Copper acetate (Cu(OAc)2), Acetic Acid Cupric Salt, Copper Acetate, Copper Acetate (Cu(C2H3O2)2), Copper Acetate (Cu(MeCO2)2), Copper Diacetate, Copper(2+) Acetate, Copper(2+) Diacetate, Copper(II) Acetate, Crystallized Verdigris, Crystals of Venus, Cupric Acetate, Cupric Diacetate, Neutral Verdigris, Ploss Blue, Venus Copper
InChI Formula InChI=1S/2C2H4O2.Cu/c2*1-2(3)4;/h2*1H3,(H,3,4);/p-2
SMILES O=C(C)[O-].O=C(C)[O-].[Cu]
IUPAC Name copper;diacetate
Molecular Weight (g/mol) 181.64
Formula Weight 180.96
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.