accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COCH3

CAS RN 142-96-1

IUPAC Name: 1-butoxybutane
Synonyms: di-N-butyl ether
Molecular Weight (g/mol): 130.23
Molecular Formula: C8H18O
InChi Key: DURPTKYDGMDSBL-UHFFFAOYSA-N
SMILES: CCCCOCCCC
Quantity 
  • (4)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (1)
  • (2)
  • (2)
  • (6)
Boiling Point 
  • (1)
  • (2)
  • (6)
  • (2)
Grade 
  • (3)
  • (3)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (6)
  • (2)
  • (4)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (6)
  • (1)
  • (2)
  • (6)
  • (2)
  • (3)
  • (3)
  • (2)
  • (1)
  • (8)