CAS RN 1421227-53-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Tocris Bioscience™ WS 6
Induces growth and proliferation of pancreatic islet cells, murine totipotent stem cells, GABAergic neurons and hippocampal neurons