missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ WS 6
Description
WS 6 induces growth and proliferation of cultured islet α and β cells in vitro (EC50 = 0.28 μM at β cells). Inhibits growth of FLT3-ITD+ AmL and K562 CmL cells (IC50 values are <0.24 nM and 40.48 nM, respectively). Reduces blood glucose in an animal model of type 1 diabetes. In combination with TTNPB (Cat. No. 0761) and 1-azakenpaullone, WS 6 induces and maintains murine totipotent stem cells. In an animal model of depression WS 6 induces dendrite complexity, enhances hippocampal neurogenesis, increases GABAergic neurons and GABA expression, and displays antidepressant and anxiolytic activity. WS 6 is predicted to be a receptor tyrosine kinase inhibitor.
Specifications
Specifications
| Chemical Name or Material | 4-[[6-[(Cyclopropylcarbonyl)amino]-4-pyrimidinyl]oxy]-N-[4-[(4-methyl-1-piperazinyl)methyl]-3-(trifluoromethyl)phenyl]benzeneacetamide |
| CAS | 1421227-53-3 |
| Quantity | 10 mg |
| Target | Compounds for Stem Cell Proliferation and Renewal |
| Molecular Formula | C29H31F3N6O3 |
| Purity | 98% |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?