accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CCH3O(E)

CAS RN 14309-57-0

IUPAC Name: (3E)-non-3-en-2-one
Synonyms: (3E)-non-3-en-2-one
Molecular Weight (g/mol): 140.23
Molecular Formula: C9H16O
InChi Key: HDKLIZDXVUCLHQ-BQYQJAHWSA-N
SMILES: CCCCC\C=C\C(C)=O
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results