CAS RN 14319-13-2
IUPAC Name:
tris(5-hydroxy-2,2,6,6-tetramethylhept-4-en-3-one) lanthanum
Synonyms:
tris(5-hydroxy-2,2,6,6-tetramethylhept-4-en-3-one) lanthanum
Molecular Weight (g/mol):
691.74
Molecular Formula:
C33H60LaO6
InChi Key:
VTNJXVDFRGBYJC-UHFFFAOYSA-N
SMILES:
[La].CC(C)(C)C(O)=CC(=O)C(C)(C)C.CC(C)(C)C(O)=CC(=O)C(C)(C)C.CC(C)(C)C(O)=CC(=O)C(C)(C)C
1
–
1
of
1
results
1
Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)lanthanum(III), 98%
CAS: 14319-13-2 Molecular Formula: C33H60LaO6 Molecular Weight (g/mol): 691.742 MDL Number: MFCD00010466 InChI Key: VTNJXVDFRGBYJC-LWTKGLMZSA-N Synonym: Lanthanum(III)-DPM; La(TMHD)3 PubChem CID: 72376397 IUPAC Name: (Z)-5-hydroxy-2,2,6,6-tetramethylhept-4-en-3-one;lanthanum SMILES: CC(C)(C)C(=CC(=O)C(C)(C)C)O.CC(C)(C)C(=CC(=O)C(C)(C)C)O.CC(C)(C)C(=CC(=O)C(C)(C)C)O.[La]