accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 143262-10-6

H3CCH3H3COON

CAS RN 143262-10-6

IUPAC Name: tert-butyl 2,3-dihydro-1H-indole-1-carboxylate
Synonyms: tert-butyl 2,3-dihydroindole-1-carboxylate
Molecular Weight (g/mol): 219.28
Molecular Formula: C13H17NO2
InChi Key: GWAXLDLPPZPQLO-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)N1CCC2=CC=CC=C12
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results