accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 143262-20-8

H3CCH3H3COONOOH

CAS RN 143262-20-8

IUPAC Name: 1-[(tert-butoxy)carbonyl]-2,3-dihydro-1H-indole-7-carboxylic acid
Synonyms: 1-(tert-butoxycarbonyl)-2,3-dihydroindole-7-carboxylic acid
Molecular Weight (g/mol): 263.29
Molecular Formula: C14H17NO4
InChi Key: SUAMIYWLXFROHE-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)N1CCC2=CC=CC(C(O)=O)=C12
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results