accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 143418-49-9

HOBOHFFF

CAS RN 143418-49-9

IUPAC Name: (3,4,5-trifluorophenyl)boronic acid
Synonyms: 3,4,5-trifluorophenylboronic acid
Molecular Weight (g/mol): 175.9
Molecular Formula: C6H4BF3O2
InChi Key: UHDDEIOYXFXNNJ-UHFFFAOYSA-N
SMILES: OB(O)C1=CC(F)=C(F)C(F)=C1
Percent Purity 
  • (1)
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (3)