missing translation for 'onlineSavingsMsg'
Learn More

3,4,5-Trifluorobenzeneboronic acid, 97%

Product Code. 11395025
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11395025 1 g 1g
11305035 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11395025 Supplier Thermo Scientific Alfa Aesar Supplier No. L18519.03
 Restricted Item
Estimated Shipment: 18-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 143418-49-9 | C6H4BF3O2 | 175.901 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 143418-49-9
Molecular Formula C6H4BF3O2
Molecular Weight (g/mol) 175.901
MDL Number MFCD02093069
InChI Key UHDDEIOYXFXNNJ-UHFFFAOYSA-N
Synonym 3,4,5-trifluorophenyl boronic acid, 3,4,5-trifluorobenzeneboronic acid, 3,4,5-trifluorophenyl boranediol, boronic acid, b-3,4,5-trifluorophenyl, 3,4,5-trifluoro phenylboric acid, boronic acid, 3,4,5-trifluorophenyl, 3,4,5-trifluorophenylboronicacid, pubchem2340, acmc-209cqj
PubChem CID 2734671
IUPAC Name (3,4,5-trifluorophenyl)boronic acid
SMILES B(C1=CC(=C(C(=C1)F)F)F)(O)O

Specifications

Melting Point 275°C to 279°C
Quantity 1 g
Beilstein 7371914
Formula Weight 175.9
Percent Purity 97%
Chemical Name or Material 3,4,5-Trifluorobenzeneboronic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.