accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 14447-18-8

OON

CAS RN 14447-18-8

IUPAC Name: benzyl 2-cyanoacetate
Synonyms: benzyl 2-cyanoacetate
Molecular Weight (g/mol): 175.19
Molecular Formula: C10H9NO2
InChi Key: RCUIWQWWDLZNMS-UHFFFAOYSA-N
SMILES: O=C(CC#N)OCC1=CC=CC=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results