missing translation for 'onlineSavingsMsg'
Learn More

Benzyl cyanoacetate, 97%

Product Code. 11356936
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11356936 100 g 100g
11346936 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11356936 Supplier Thermo Scientific Alfa Aesar Supplier No. L06644.22
 Restricted Item
Estimated Shipment: 14-10-2026
Add to basket
This item is not returnable. View return policy

CAS: 14447-18-8 | C10H9NO2 | 175.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14447-18-8
Molecular Formula C10H9NO2
Molecular Weight (g/mol) 175.19
MDL Number MFCD00051719
InChI Key RCUIWQWWDLZNMS-UHFFFAOYSA-N
Synonym benzyl cyanoacetate, acetic acid, cyano-, phenylmethyl ester, cyanoacetic acid benzyl ester, benzyl-cyanoacetate, acetic acid, cyano-, benzyl ester, acmc-20alph, phenylmethyl 2-cyanoacetate, cyano-acetic acid benzyl ester, benzyl ester of cyanoacetic acid
PubChem CID 560818
IUPAC Name benzyl 2-cyanoacetate
SMILES O=C(CC#N)OCC1=CC=CC=C1

Specifications

Melting Point 26°C to 30°C
Boiling Point 140°C (0.8 mmHg)
Flash Point >100°C (212°F)
Odor Odorless
Refractive Index 1.5195
Quantity 100 g
Beilstein 2096578
Formula Weight 175.18
Percent Purity 97%
Chemical Name or Material Benzyl cyanoacetate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.