CAS RN 1445-91-6
IUPAC Name:
(1S)-1-phenylethan-1-ol
Synonyms:
(S)-1-phenylethanol (S)-α-methylbenzenemethanol (S)-1-phenethyl alcohol
Molecular Weight (g/mol):
122.17
Molecular Formula:
C8H10O
InChi Key:
WAPNOHKVXSQRPX-ZETCQYMHSA-N
SMILES:
C[C@H](O)C1=CC=CC=C1
1
–
2
of
2
results
1
(S)-(-)-sec-Phenethyl alcohol, 99%
CAS: 1445-91-6 Molecular Formula: C8H10O Molecular Weight (g/mol): 122.17 MDL Number: MFCD00064264 InChI Key: WAPNOHKVXSQRPX-ZETCQYMHSA-N Synonym: s-1-phenylethanol,s---1-phenylethanol,1s-1-phenylethanol,1s-1-phenylethan-1-ol,unii-2mic4qly2x,1-phenylethanol, s,2mic4qly2x,s-1-phenethyl alcohol,s-1-phenyl-1-ethanol,s-1-phenylethan-1-ol PubChem CID: 443135 ChEBI: CHEBI:16346 SMILES: C[C@H](O)C1=CC=CC=C1
2
(S)-(-)-1-Phenylethanol, 97+%
CAS: 1445-91-6 Molecular Formula: C8H10O Molecular Weight (g/mol): 122.17 MDL Number: MFCD00064264 InChI Key: WAPNOHKVXSQRPX-ZETCQYMHSA-N PubChem CID: 443135 ChEBI: CHEBI:16346 SMILES: C[C@H](O)C1=CC=CC=C1