accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NiOOOCCCH3CH3H3CH3CCH3H3CH3CCH3H3CCH3CH3H3CHHO

CAS RN 14481-08-4

IUPAC Name: bis(5-hydroxy-2,2,6,6-tetramethylhept-4-en-3-one) nickel
Synonyms: bis(5-hydroxy-2,2,6,6-tetramethylhept-4-en-3-one) nickel
Molecular Weight (g/mol): 427.25
Molecular Formula: C22H40NiO4
InChi Key: LTUQBPCIVSGVNZ-UHFFFAOYSA-N
SMILES: [Ni].CC(C)(C)C(O)=CC(=O)C(C)(C)C.CC(C)(C)C(O)=CC(=O)C(C)(C)C
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results