CAS RN 1449133-22-5
IUPAC Name:
{3-fluoro-4-[(pyridin-2-yl)carbamoyl]phenyl}boronic acid
Synonyms:
3-fluoro-4-[(pyridin-2-yl)carbamoyl]phenylboronic acid
Molecular Weight (g/mol):
260.03
Molecular Formula:
C12H10BFN2O3
InChi Key:
CIQKUPRZPUUGGK-UHFFFAOYSA-N
SMILES:
OB(O)C1=CC(F)=C(C=C1)C(=O)NC1=CC=CC=N1
1
–
1
of
1
results
1
3-Fluoro-4-(2-pyridylcarbamoyl)benzeneboronic acid, 97%
CAS: 1449133-22-5 Molecular Formula: C12H10BFN2O3 Molecular Weight (g/mol): 260.031 MDL Number: MFCD20265181 InChI Key: CIQKUPRZPUUGGK-UHFFFAOYSA-N Synonym: 3-fluoro-4-2-pyridylcarbamoyl benzeneboronic acid,3-fluoro-4-pyridin-2-yl carbamoyl phenylboronic acid,3-fluoro-4-2-pyridylcarbamoyl phenylboronic acid PubChem CID: 73995642 IUPAC Name: [3-fluoro-4-(pyridin-2-ylcarbamoyl)phenyl]boronic acid SMILES: B(C1=CC(=C(C=C1)C(=O)NC2=CC=CC=N2)F)(O)O