missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoro-4-(2-pyridylcarbamoyl)benzeneboronic acid, 97%

Product Code. 15420796
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15420796 250 mg 250mg
15410796 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15420796 Supplier Thermo Scientific Alfa Aesar Supplier No. H54122.MD
 Restricted Item
Estimated Shipment: 15-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1449133-22-5 | C12H10BFN2O3 | 260.031 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1449133-22-5
Molecular Formula C12H10BFN2O3
Molecular Weight (g/mol) 260.031
MDL Number MFCD20265181
InChI Key CIQKUPRZPUUGGK-UHFFFAOYSA-N
Synonym 3-fluoro-4-2-pyridylcarbamoyl benzeneboronic acid, 3-fluoro-4-pyridin-2-yl carbamoyl phenylboronic acid, 3-fluoro-4-2-pyridylcarbamoyl phenylboronic acid
PubChem CID 73995642
IUPAC Name [3-fluoro-4-(pyridin-2-ylcarbamoyl)phenyl]boronic acid
SMILES B(C1=CC(=C(C=C1)C(=O)NC2=CC=CC=N2)F)(O)O

Specifications

Quantity 250 mg
Formula Weight 260.03
Percent Purity 97%
Chemical Name or Material 3-Fluoro-4-(2-pyridylcarbamoyl)benzeneboronic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.