accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CO(S)(S)(S)(S)OH(R)H3C(S)(S)H3C

CAS RN 145-13-1

IUPAC Name: 1-[(1S,3aS,3bS,7S,9aR,9bS,11aS)-7-hydroxy-9a,11a-dimethyl-1H,2H,3H,3aH,3bH,4H,6H,7H,8H,9H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-1-yl]ethan-1-one
Synonyms: pregnenolone pregnenolona enelone pregnetan pregneton skinostelon pregn-5-ene-3b-ol-20-one prenolon regnosone delta5-pregnenolone
Molecular Weight (g/mol): 316.49
Molecular Formula: C21H32O2
InChi Key: ORNBQBCIOKFOEO-QGVNFLHTSA-N
SMILES: CC(=O)[C@H]1CC[C@H]2[C@@H]3CC=C4C[C@@H](O)CC[C@]4(C)[C@H]3CC[C@]12C
Molecular Weight (g/mol) 
  • (3)
  • (7)
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
Certifications/Compliance 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results