accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NH2OHONH2OHO

CAS RN 145-49-3

IUPAC Name: 1,5-diamino-4,8-dihydroxy-9,10-dihydroanthracene-9,10-dione
Synonyms: diaminoanthrarufin
Molecular Weight (g/mol): 270.24
Molecular Formula: C14H10N2O4
InChi Key: HSYLKWSCFRLSKB-UHFFFAOYSA-N
SMILES: NC1=CC=C(O)C2=C1C(=O)C1=C(O)C=CC(N)=C1C2=O
Grade 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results