accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 1450-63-1

IUPAC Name: (1,4,4-triphenylbuta-1,3-dien-1-yl)benzene
Synonyms: tetraphenyl butadiene
Molecular Weight (g/mol): 358.48
Molecular Formula: C28H22
InChi Key: KLCLIOISYBHYDZ-UHFFFAOYSA-N
SMILES: C(C=C(C1=CC=CC=C1)C1=CC=CC=C1)=C(C1=CC=CC=C1)C1=CC=CC=C1
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
Physical Form 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results