accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3NNH3CFFFSOOO

CAS RN 145022-44-2

IUPAC Name: 1-ethyl-3-methyl-1H-imidazol-3-ium trifluoromethanesulfonate
Synonyms: 1-ethyl-3-methylimidazolium triflate
Molecular Weight (g/mol): 260.23
Molecular Formula: C7H11F3N2O3S
InChi Key: ZPTRYWVRCNOTAS-UHFFFAOYSA-M
SMILES: CCN1C=C[N+](C)=C1.[O-]S(=O)(=O)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results