missing translation for 'onlineSavingsMsg'
Learn More

1-Ethyl-3-methylimidazolium trifluoromethanesulfonate, 98+%

Product Code. 11319715
Change view
Click to view available options
Quantity:
2 g
10 g
Unit Size:
10g
2g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11319715 10 g 10g
11309715 2 g 2g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11319715 Supplier Thermo Scientific Alfa Aesar Supplier No. L19764.09
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 145022-44-2 | C7H11F3N2O3S | 260.231 g/mol

Recently used in the preparation of hydrophobic, highly conductive ambient-temperature molten salts, and in the preparation of dual intercalating molten electrolyte batteries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Recently used in the preparation of hydrophobic, highly conductive ambient-temperature molten salts, and in the preparation of dual intercalating molten electrolyte batteries.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, water, moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 145022-44-2
Molecular Formula C7H11F3N2O3S
Molecular Weight (g/mol) 260.231
MDL Number MFCD00210008
InChI Key ZPTRYWVRCNOTAS-UHFFFAOYSA-M
Synonym 1-ethyl-3-methylimidazolium trifluoromethanesulfonate, 1-ethyl-3-methylimidazolium triflate, 1-ethyl-3-methyl-1h-imidazol-3-ium trifluoromethanesulfonate, acmc-209ctv, dsstox_cid_27941, dsstox_rid_82693, dsstox_gsid_47965, 1-ethyl-3-methylimidazoliumtrifluoromethanesulfonate, ksc685i3d, 1-ethyl-3-methylimidazolium trifluoromethanesulfon
PubChem CID 2758874
IUPAC Name 1-ethyl-3-methylimidazol-3-ium;trifluoromethanesulfonate
SMILES CCN1C=C[N+](=C1)C.C(F)(F)(F)S(=O)(=O)[O-]

Specifications

Density 1.387
Flash Point None
Refractive Index 1.435
Quantity 10 g
Sensitivity Hygroscopic
Solubility Information Soluble in water.
Formula Weight 260.24
Percent Purity ≥98%
Chemical Name or Material 1-Ethyl-3-methylimidazolium trifluoromethanesulfonate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.