CAS RN 145356-32-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
4-Hydroxy Xylazine, TRC
CAS: 145356-32-7 Molecular Formula: C12 H16 N2 O S Molecular Weight (g/mol): 236.33 IUPAC Name: 4-(5,6-dihydro-4H-1,3-thiazin-2-ylamino)-3,5-dimethylphenol SMILES: Cc1cc(O)cc(C)c1NC2=NCCCS2