missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxy Xylazine, TRC

Product Code. 30135937
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30135937 10 mg 10mg
30003891 5 mg 5mg
30011713 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30135937 Supplier Toronto Research Chemicals Supplier No. TRCH99630010MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 145356-32-7
Molecular Formula C12 H16 N2 O S
Molecular Weight (g/mol) 236.33
IUPAC Name 4-(5,6-dihydro-4H-1,3-thiazin-2-ylamino)-3,5-dimethylphenol
SMILES Cc1cc(O)cc(C)c1NC2=NCCCS2

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C12H16N2OS/c1-8-6-10(15)7-9(2)11(8)14-12-13-4-3-5-16-12/h6-7,15H,3-5H2,1-2H3,(H,13,14)
Formula Weight 236.0983
Percent Purity >95%
Chemical Name or Material 4-Hydroxy Xylazine
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.