accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 14542-12-2

SNHO

CAS RN 14542-12-2

IUPAC Name: (1,3-thiazol-2-yl)methanol
Synonyms: 1,3-thiazol-2-ylmethanol
Molecular Weight (g/mol): 115.15
Molecular Formula: C4H5NOS
InChi Key: JNHDLNXNYPLBMJ-UHFFFAOYSA-N
SMILES: OCC1=NC=CS1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results