accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CO2H

CAS RN 1455-13-6

IUPAC Name: methan(²H)ol
Synonyms: methan(²H)ol
Molecular Weight (g/mol): 33.05
Molecular Formula: CH4O
InChi Key: OKKJLVBELUTLKV-VMNATFBRSA-N
SMILES: [2H]OC
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)