accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OH(R)(R)HO

CAS RN 1460-57-7

IUPAC Name: (1R,2R)-cyclohexane-1,2-diol
Synonyms: trans-cyclohexane-1,2-diol trans-1,2-dihydroxycyclohexane
Molecular Weight (g/mol): 116.16
Molecular Formula: C6H12O2
InChi Key: PFURGBBHAOXLIO-PHDIDXHHSA-N
SMILES: O[C@@H]1CCCC[C@H]1O
Quantity 
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
Boiling Point 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)