accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 146137-76-0

IUPAC Name: 2-fluoro-5-iodobenzaldehyde
Synonyms: 2-fluoro-5-iodobenzaldehyde
Molecular Weight (g/mol): 250.01
Molecular Formula: C7H4FIO
InChi Key: BIRCCQCPGMMGPJ-UHFFFAOYSA-N
SMILES: FC1=C(C=O)C=C(I)C=C1
Percent Purity 
  • (3)
  • (1)
Quantity 
  • (2)
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results