missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-5-iodobenzaldehyde, 97%, Thermo Scientific™

Product Code. 10193334 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10193334 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10193334 Supplier Thermo Scientific Acros Supplier No. 399990010

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 146137-76-0
CAS Min % 96.0
CAS Max % 100.0
Melting Point 45°C to 48°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Molecular Formula C7H4FIO
Linear Formula FC6H3(I)CHO
MDL Number MFCD03093999
Quantity 1 g
Synonym 2-fluoro-5-iodo-benzaldehyde, pubchem4222, acmc-1bypb, c7h4flo, 6-fluoro-3-iodobenzaldehyde, 2-fluoranyl-5-iodanyl-benzaldehyde, 2-fluoro-5-iodobenzaldehyde
InChI Key BIRCCQCPGMMGPJ-UHFFFAOYSA-N
SMILES FC1=C(C=O)C=C(I)C=C1
IUPAC Name 2-fluoro-5-iodobenzaldehyde
Molecular Weight (g/mol) 250.01
PubChem CID 16217058
Formula Weight 250.01
Percent Purity 97%
Physical Form Crystals and/or Chunks
Chemical Name or Material 2-Fluoro-5-iodobenzaldehyde
Show More Show Less
compliance-icons
Product Identifier
  • 2-Fluoro-5-iodobenzaldehyde
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.