accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 1462-03-9

CH3HO

CAS RN 1462-03-9

IUPAC Name: 1-methylcyclopentan-1-ol
Synonyms: 1-hydroxy-1-methylcyclopentane
Molecular Weight (g/mol): 100.16
Molecular Formula: C6H12O
InChi Key: CAKWRXVKWGUISE-UHFFFAOYSA-N
SMILES: CC1(O)CCCC1
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results