accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

GdOOOOOOCCCCH3H3CH3CCH3CH3CH3CH3H3CCH3H3CH3CCH3CH3CH3H3CH3CCH3H3CHHH

CAS RN 14768-15-1

IUPAC Name: tris(5-hydroxy-2,2,6,6-tetramethylhept-4-en-3-one) gadolinium
Synonyms: tris(5-hydroxy-2,2,6,6-tetramethylhept-4-en-3-one) gadolinium
Molecular Weight (g/mol): 710.09
Molecular Formula: C33H60GdO6
InChi Key: KUKZOIVDRFKQPE-UHFFFAOYSA-N
SMILES: [Gd].CC(C)(C)C(O)=CC(=O)C(C)(C)C.CC(C)(C)C(O)=CC(=O)C(C)(C)C.CC(C)(C)C(O)=CC(=O)C(C)(C)C
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results