accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OF

CAS RN 1481-32-9

IUPAC Name: 6-fluoro-2,3-dihydro-1H-inden-1-one
Synonyms: 6-fluoro-2,3-dihydroinden-1-one
Molecular Weight (g/mol): 150.15
Molecular Formula: C9H7FO
InChi Key: LVUUCFIQQHEFEJ-UHFFFAOYSA-N
SMILES: FC1=CC2=C(CCC2=O)C=C1
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results