accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3OOH3COH3C

CAS RN 149-73-5

IUPAC Name: trimethoxymethane
Synonyms: trimethyl orthoformate
Molecular Weight (g/mol): 106.12
Molecular Formula: C4H10O3
InChi Key: PYOKUURKVVELLB-UHFFFAOYSA-N
SMILES: COC(OC)OC
Molecular Weight (g/mol) 
  • (8)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
Boiling Point 
  • (3)
  • (6)
Percent Purity 
  • (7)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (3)
  • (8)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)
  • (6)
  • (7)
  • (2)
  • (4)