accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COBOHHO

CAS RN 149104-90-5

IUPAC Name: (4-acetylphenyl)boronic acid
Synonyms: 4-acetylphenylboronic acid
Molecular Weight (g/mol): 163.97
Molecular Formula: C8H9BO3
InChi Key: OBQRODBYVNIZJU-UHFFFAOYSA-N
SMILES: CC(=O)C1=CC=C(C=C1)B(O)O
Quantity 
  • (3)
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results