accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 1497-17-2

H3COHNO

CAS RN 1497-17-2

IUPAC Name: 3-methyl-3-phenylpyrrolidine-2,5-dione
Synonyms: 3-methyl-3-phenylpyrrolidine-2,5-dione
Molecular Weight (g/mol): 189.21
Molecular Formula: C11H11NO2
InChi Key: UDESUGJZUFALAM-UHFFFAOYNA-N
SMILES: CC1(CC(=O)NC1=O)C1=CC=CC=C1
Quantity 
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results