missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-2-phenylsuccinimide, 99%

Product Code. 15437927
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15437927 5 g 5g
15427927 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15437927 Supplier Thermo Scientific Alfa Aesar Supplier No. H51038.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1497-17-2 | C11H11NO2 | 189.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identificatori chimici

CAS 1497-17-2
Molecular Formula C11H11NO2
Molecular Weight (g/mol) 189.21
MDL Number MFCD00005496
InChI Key UDESUGJZUFALAM-UHFFFAOYNA-N
Synonym 2-methyl-2-phenylsuccinimide, n-desmethylmethsuximide, normesuximide, a-methyl-a-phenylsuccinimide, n-demethylmethsuximide, 2,5-pyrrolidinedione, 3-methyl-3-phenyl, alpha-methyl-alpha-phenylsuccinimide, 3-methyl-3-phenyl-2,5-pyrrolidinedione, .alpha.-methyl-.alpha.-phenylsuccinimide, normethsuximide
PubChem CID 92154
IUPAC Name 3-methyl-3-phenylpyrrolidine-2,5-dione
SMILES CC1(CC(=O)NC1=O)C1=CC=CC=C1

Specifica

Quantity 5 g
Formula Weight 189.21
Percent Purity 99%
Chemical Name or Material 2-Methyl-2-phenylsuccinimide

RUO – Research Use Only

Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.