accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

BrON

CAS RN 149806-06-4

IUPAC Name: 6-bromopyridine-3-carbaldehyde
Synonyms: 6-bromopyridine-3-carbaldehyde
Molecular Weight (g/mol): 186.01
Molecular Formula: C6H4BrNO
InChi Key: PVUKGNBRJFTFNJ-UHFFFAOYSA-N
SMILES: BrC1=CC=C(C=O)C=N1
Quantity 
  • (3)
  • (1)
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (2)
  • (3)
  • (3)