accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 150-50-5

IUPAC Name: tris(butylsulfanyl)phosphane
Synonyms: merphos
Molecular Weight (g/mol): 298.5
Molecular Formula: C12H27PS3
InChi Key: KLAPGAOQRZTCBI-UHFFFAOYSA-N
SMILES: CCCCSP(SCCCC)SCCCC
Molecular Weight (g/mol) 
  • (1)
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
Solvent or Matrix 
  • (1)
  • (1)
Concentration 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (3)
  • (1)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)