missing translation for 'onlineSavingsMsg'
Learn More

Merphos, TRC

Product Code. 30134046
Change view
Click to view available options
Quantity:
5 mg
25 mg
50 mg
Unit Size:
25mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30134046 50 mg 50mg
30005753 25 mg 25mg
30006252 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30134046 Supplier Toronto Research Chemicals Supplier No. TRCM25770050MG
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 150-50-5
Molecular Formula C12 H27 P S3
Molecular Weight (g/mol) 298.51
Synonym Butyl phosphorotrithioite (6CI), Butyl phosphorotrithioite ((BuS)3P), Folex, Merphos, NSC 27720, Tributyl phosphorotrithioite, Tributyl trithiophosphite, Tris(butylthio)phosphine
IUPAC Name tris(butylsulfanyl)phosphane
SMILES CCCCSP(SCCCC)SCCCC

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C12H27PS3/c1-4-7-10-14-13(15-11-8-5-2)16-12-9-6-3/h4-12H2,1-3H3
Formula Weight 298.1012
Chemical Name or Material Merphos
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.